Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ubxn2a
Entrez Gene ID
217379
Gene Name
UBX domain protein 2A
Gene Aliases
6330407P03Rik, Ubxd4
Chromosome Location
Chr.12: 4876647 - 4907781 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145441.3 NP_663416.1 6 - 7 773 113
GenBank mRNA AK031821.1 - 6 - 7 774 113
AK041556.1 - 6 - 7 810 113
BC004632.1 - 6 - 7 725 113

View Full Product Details