Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Atad2b
Entrez Gene ID
320817
Gene Name
ATPase family, AAA domain containing 2B
Gene Aliases
1110014E10Rik, BC032887, C79189, D530031C13Rik
Chromosome Location
Chr.12: 4916084 - 5047410 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099628.1 NP_001093098.1 11 - 12 1654 105

View Full Product Details