The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024703.1 | NP_001019874.1 | 15 - 16 | 2082 | 150 |
GenBank mRNA | BC086658.1 | - | 15 - 16 | 2082 | 150 |