Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ctbs
Entrez Gene ID
74245
Gene Name
chitobiase, di-N-acetyl-
Gene Aliases
2210401K11Rik, CTB
Chromosome Location
Chr.3: 146450467 - 146465849 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293672.1 NP_001280601.1 5 - 6 776 78
GenBank mRNA BC022594.1 - 4 - 5 537 78
DQ100384.1 - 5 - 6 750 78

View Full Product Details