Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rundc3b
Entrez Gene ID
242819
Gene Name
RUN domain containing 3B
Gene Aliases
Gm440
Chromosome Location
Chr.5: 8490336 - 8623027 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198620.1 NP_941022.1 7 - 8 1141 70
GenBank mRNA BC057375.1 - 3 - 4 354 70
BC058808.1 - 7 - 8 1141 70

View Full Product Details