Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mphosph10
Entrez Gene ID
67973
Gene Name
M-phase phosphoprotein 10 (U3 small nucleolar ribonucleoprotein)
Gene Aliases
2810453H10Rik, 5730405D16Rik, AI326008
Chromosome Location
Chr.7: 64376541 - 64392236 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026483.2 NP_080759.2 2 - 3 98
GenBank mRNA BC018177.1 - 2 - 3 98
BC038033.1 - 2 - 3 98
BC049270.1 - 2 - 3 791 98
CK390971.1 - 1 - 2 98

View Full Product Details