Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prss52
Entrez Gene ID
73382
Gene Name
protease, serine 52
Gene Aliases
1700049K14Rik, Tesp3
Chromosome Location
Chr.14: 64104323 - 64113751 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028525.2 NP_082801.2 2 - 3 337 69
GenBank mRNA AB009660.1 - 2 - 3 374 69
AK006746.1 - 2 - 3 336 69
BC049714.1 - 2 - 3 337 69

View Full Product Details