The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193631.1 | NP_001180560.1 | 3 - 4 | 509 | 63 |
GenBank mRNA | AK005740.1 | - | 3 - 4 | 514 | 63 |
BC048657.1 | - | 3 - 4 | 509 | 63 |