The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145837.3 | NP_665836.2 | 2 - 3 | 388 | 69 |
GenBank mRNA | AF458063.1 | - | 1 - 2 | 310 | 69 |
AK131917.1 | - | 2 - 3 | 437 | 69 |