The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199020.1 | NP_001185949.1 | 8 - 9 | 1383 | 61 |
NM_021421.4 | NP_067396.3 | 7 - 8 | 1518 | 61 | |
NR_037578.1 | 7 - 8 | 1477 | 61 | ||
NR_037579.1 | 6 - 7 | 1192 | 61 | ||
GenBank mRNA | AB041602.1 | - | 5 - 6 | 1051 | 61 |
AK077039.1 | - | 7 - 8 | 1465 | 61 | |
AK088834.1 | - | 6 - 7 | 1151 | 61 | |
AK135709.1 | - | 8 - 9 | 1380 | 61 | |
AK158731.1 | - | 7 - 8 | 1534 | 61 | |
AK166619.1 | - | 8 - 9 | 1382 | 61 | |
AK169653.1 | - | 6 - 7 | 1102 | 61 | |
BC024907.1 | - | 7 - 8 | 1478 | 61 |