Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Disp1
Entrez Gene ID
68897
Gene Name
dispatched homolog 1 (Drosophila)
Gene Aliases
1190008H24Rik, DispA
Chromosome Location
Chr.1: 183086264 - 183221536 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278218.1 NP_001265147.1 3 - 4 863 59
NM_001278219.1 NP_001265148.1 3 - 4 823 59
NM_001278220.1 NP_001265149.1 2 - 3 706 59
NM_026866.3 NP_081142.3 4 - 5 951 59
GenBank mRNA AK004521.1 - 2 - 3 681 59
AK138276.1 - 4 - 5 908 59
AK170113.1 - 3 - 4 818 59
AY144589.1 - 2 - 3 661 59
BC043102.1 - 3 - 4 824 59
BC059225.1 - 3 - 4 824 59

View Full Product Details