Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psmd6
Entrez Gene ID
66413
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 6
Gene Aliases
2400006A19Rik
Chromosome Location
Chr.14: 14112185 - 14120904 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025550.2 NP_079826.2 5 - 6 851 118
GenBank mRNA AJ252147.1 - 5 - 6 887 118
AK002343.1 - 5 - 6 887 118
AK010287.1 - 5 - 6 884 118
AK211149.1 - 2 - 3 129 117
BC006869.1 - 5 - 6 851 118

View Full Product Details