Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psmd6
Entrez Gene ID
66413
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 6
Gene Aliases
2400006A19Rik
Chromosome Location
Chr.14: 14112185 - 14120904 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025550.2 NP_079826.2 6 - 7 1017 59
GenBank mRNA AJ252147.1 - 6 - 7 1053 59
AK002343.1 - 6 - 7 1053 59
AK010287.1 - 6 - 7 1050 59
AK178742.1 - 3 - 4 296 59
AK206699.1 - 1 - 2 137 59
BC006869.1 - 6 - 7 1017 59

View Full Product Details