Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fgf17
Entrez Gene ID
14171
Gene Name
fibroblast growth factor 17
Gene Aliases
-
Chromosome Location
Chr.14: 70636203 - 70643014 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008004.4 NP_032030.1 1 - 2 546 145
GenBank mRNA AK077555.1 - 1 - 2 548 145
AK082858.1 - 1 - 2 437 145
BC119033.1 - 1 - 2 532 145

View Full Product Details