Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ocrl
Entrez Gene ID
320634
Gene Name
oculocerebrorenal syndrome of Lowe
Gene Aliases
9530014D17Rik, BB143339, OCRL1
Chromosome Location
Chr.X: 47912456 - 47966428 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177215.3 NP_796189.2 11 - 12 1249 93
GenBank mRNA AK035315.1 - 11 - 12 1252 93
AK041963.1 - 11 - 12 1251 93
AK044320.1 - 10 - 11 1171 93
AK047906.1 - 11 - 12 1268 93
BC068146.1 - 11 - 12 1249 93

View Full Product Details