Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dmpk
Entrez Gene ID
13400
Gene Name
dystrophia myotonica-protein kinase
Gene Aliases
DM, DMK, Dm15, MDPK, MT-PK
Chromosome Location
Chr.7: 19083834 - 19093821 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190490.1 NP_001177419.1 15 - 16 1813 74
NM_032418.2 NP_115794.1 14 - 15 1891 74
GenBank mRNA BC056615.1 - 14 - 15 1980 74
BC075715.1 - 15 - 16 1721 74
S60313.1 - 10 - 11 1565 74
S60314.1 - 11 - 12 1394 74
S60315.1 - 11 - 12 1607 74
S60316.1 - 10 - 11 1268 74

View Full Product Details