Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dmpk
Entrez Gene ID
13400
Gene Name
dystrophia myotonica-protein kinase
Gene Aliases
DM, DMK, Dm15, MDPK, MT-PK
Chromosome Location
Chr.7: 19083834 - 19093821 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190490.1 NP_001177419.1 5 - 6 727 87
NM_001190491.1 NP_001177420.1 5 - 6 727 87
NM_032418.2 NP_115794.1 5 - 6 727 87
GenBank mRNA BC024150.1 - 5 - 6 714 87
BC056615.1 - 5 - 6 706 87
BC075715.1 - 5 - 6 635 87
S60313.1 - 2 - 3 234 87
S60314.1 - 2 - 3 234 87
S60315.1 - 2 - 3 234 87
S60316.1 - 2 - 3 234 87

View Full Product Details