Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Styx
- Entrez Gene ID
-
56291
- Gene Name
- serine/threonine/tyrosine interaction protein
- Gene Aliases
-
0610039A20Rik, hStyxb
- Chromosome Location
-
Chr.14: 45351186 - 45376884 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK002822.1 |
- |
- |
980 |
95 |
View Full Product Details