The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170552.1 | NP_001164023.1 | 4 - 5 | 713 | 141 |
NM_030675.3 | NP_109600.2 | 4 - 5 | 667 | 141 | |
NR_033173.1 | 4 - 5 | 766 | 141 | ||
GenBank mRNA | AF306509.1 | - | 4 - 5 | 380 | 141 |
AF310134.1 | - | 4 - 5 | 643 | 141 | |
AK039264.1 | - | - | 688 | 141 | |
AK144667.1 | - | 4 - 5 | 649 | 141 | |
AK172215.1 | - | 2 - 3 | 4350 | 141 | |
AY328895.1 | - | 3 - 4 | 349 | 141 | |
AY464945.1 | - | 3 - 4 | 349 | 141 | |
BC016087.1 | - | 4 - 5 | 617 | 141 | |
BC044820.1 | - | 3 - 4 | 408 | 141 | |
BC054819.1 | - | 4 - 5 | 758 | 141 | |
BC085252.1 | - | 4 - 5 | 473 | 141 | |
BC141042.1 | - | 4 - 5 | 511 | 141 |