The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161731.2 | NP_001155203.1 | 1 - 2 | 179 | 55 |
GenBank mRNA | AK171251.1 | - | 1 - 2 | 107 | 55 |
BF721354.1 | - | 1 - 2 | 52 | 55 |