The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_037579.1 | 1 - 2 | 250 | 106 | |
GenBank mRNA | AB041602.1 | - | 1 - 2 | 236 | 106 |
AK169653.1 | - | 1 - 2 | 160 | 106 |