Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ebf2
Entrez Gene ID
13592
Gene Name
early B cell factor 2
Gene Aliases
D14Ggc1e, EBF-2, Mmot1, O/E-3, OE-3
Chromosome Location
Chr.14: 67233292 - 67430952 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276387.1 NP_001263316.1 8 - 9 1604 75
NM_010095.6 NP_034225.1 8 - 9 1599 75
GenBank mRNA AK053013.1 - 8 - 9 1600 75
AK139262.1 - 8 - 9 1311 75
BC049188.1 - 7 - 8 898 75
BC050922.1 - 7 - 8 1121 75
U71189.1 - 7 - 8 784 75
U82441.1 - 7 - 8 653 75
U92703.1 - 7 - 8 1004 75

View Full Product Details