Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam73a
Entrez Gene ID
215708
Gene Name
family with sequence similarity 73, member A
Gene Aliases
A930038D06, AI852168, C030011O14Rik
Chromosome Location
Chr.3: 152273459 - 152340437 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162375.1 NP_001155847.1 12 - 13 1654 80
NM_174868.4 NP_777357.2 14 - 15 1846 80
GenBank mRNA AK045347.1 - 12 - 13 1653 80
AK083909.1 - 13 - 14 1677 80
AK085017.1 - 14 - 15 1846 80
AK143366.1 - 14 - 15 1846 80
AK166051.1 - 14 - 15 1799 80
BC098205.1 - 14 - 15 1829 80
BC138328.1 - 14 - 15 1793 80
BC138329.1 - 14 - 15 1793 80

View Full Product Details