Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tm9sf1
Entrez Gene ID
74140
Gene Name
transmembrane 9 superfamily member 1
Gene Aliases
1200014D02Rik, AI893436, MP70
Chromosome Location
Chr.14: 55635965 - 55643806 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028780.3 NP_083056.2 5 - 6 1874 72
GenBank mRNA AF269152.1 - 6 - 7 1519 72
AK004754.2 - 5 - 6 1876 72
AK090065.1 - 4 - 5 1870 72
AK152476.1 - 5 - 6 1654 72
AK152540.1 - 5 - 6 1654 72
BC007187.1 - 4 - 5 1179 72
BC017617.1 - 4 - 5 1813 72

View Full Product Details