Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc9b1
Entrez Gene ID
74446
Gene Name
solute carrier family 9, subfamily B (NHA1, cation proton antiporter 1), member 1
Gene Aliases
1700094G20Rik, 4933424B12Rik, 4933425K02Rik, Nhedc1, mtsNHE
Chromosome Location
Chr.3: 135347994 - 135397827 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028946.3 NP_083222.2 11 - 12 1882 157
GenBank mRNA AK029525.1 - 11 - 12 1883 157
EU846100.1 - 10 - 11 1413 157

View Full Product Details