The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033350.2 | NP_001028522.2 | 3 - 4 | 727 | 127 |
GenBank mRNA | AK041242.1 | - | 3 - 4 | 688 | 127 |
AK156201.1 | - | 3 - 4 | 737 | 127 | |
AY178734.1 | - | 3 - 4 | 747 | 127 | |
BC026932.1 | - | 3 - 4 | 674 | 127 | |
EU051378.1 | - | 2 - 3 | 328 | 127 |