The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033350.2 | NP_001028522.2 | 5 - 6 | 1007 | 149 |
GenBank mRNA | AK041071.1 | - | 2 - 3 | 265 | 149 |
AK041242.1 | - | 5 - 6 | 968 | 149 | |
AK156201.1 | - | 5 - 6 | 1017 | 149 | |
AY178734.1 | - | 5 - 6 | 1027 | 149 | |
EU051378.1 | - | 4 - 5 | 608 | 149 |