Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fes
Entrez Gene ID
14159
Gene Name
feline sarcoma oncogene
Gene Aliases
AI586313, BB137047, FPS, c-fes
Chromosome Location
Chr.7: 80377758 - 80387954 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010194.2 NP_034324.2 1 - 2 109 93
GenBank mRNA AK004587.1 - 1 - 2 101 93
AK155861.1 - 1 - 2 105 93
BC038130.1 - 1 - 2 123 93

View Full Product Details