Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ahctf1
Entrez Gene ID
226747
Gene Name
AT hook containing transcription factor 1
Gene Aliases
6230412P20Rik, AV011447, Elys
Chromosome Location
Chr.1: 179744894 - 179804015 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026375.2 NP_080651.2 17 - 18 2812 58
GenBank mRNA AB059278.1 - 17 - 18 2468 58
BC138343.1 - 17 - 18 2455 58
BC138344.1 - 17 - 18 2455 58

View Full Product Details