Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prc1
Entrez Gene ID
233406
Gene Name
protein regulator of cytokinesis 1
Gene Aliases
D7Ertd348e
Chromosome Location
Chr.7: 80293786 - 80316259 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285997.1 NP_001272926.1 8 - 9 1236 71
NM_001285998.1 NP_001272927.1 8 - 9 1236 71
NM_145150.3 NP_660132.2 8 - 9 1236 71
GenBank mRNA AK029140.1 - 8 - 9 1223 71
AK088576.1 - 8 - 9 1208 71
AK135778.1 - 6 - 7 861 71
BC059808.1 - 8 - 9 1181 71

View Full Product Details