The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271447.1 | NP_001258376.1 | 1 - 2 | 140 | 294 |
NM_001271448.1 | NP_001258377.1 | 1 - 2 | 140 | 139 | |
NM_001271449.1 | NP_001258378.1 | 1 - 2 | 140 | 283 | |
NM_001271450.1 | NP_001258379.1 | 1 - 2 | 140 | 283 | |
NM_001271451.1 | NP_001258380.1 | 1 - 2 | 140 | 283 | |
GenBank mRNA | AK028343.1 | - | 1 - 2 | 141 | 283 |
AK041822.1 | - | 1 - 2 | 141 | 283 | |
AK077508.1 | - | 1 - 2 | 111 | 294 | |
AK133956.1 | - | 1 - 2 | 111 | 139 | |
AK137736.1 | - | 1 - 2 | 111 | 294 | |
AK139897.1 | - | 1 - 2 | 111 | 294 | |
BC025915.1 | - | 1 - 2 | 114 | 283 |