Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Airn
- Entrez Gene ID
-
104103
- Gene Name
- antisense Igf2r RNA
- Gene Aliases
-
2810051F02Rik, 2810434M15Rik, AI256653, AI597500, AW049873, Air, B930018I07Rik, D17Ertd663e, IGF2RAS
- Chromosome Location
-
Chr.17: 12741311 - 12859884 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002853.2 |
|
1 - 2 |
120 |
72 |
GenBank mRNA |
AK012942.1 |
- |
1 - 2 |
59 |
72 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
solute carrier family 22 (organic cation transporter), member 1 |
Slc22a1 |
Lx1, Oct1, Orct, Orct1 |
-65.5kb
|
Anti-sense |
Chr. - 12648874 - 12675838 on Build GRCm38 |
Downstream |
MAS1 oncogene |
Mas1 |
Mas-1, Mgra |
23.3kb
|
Anti-sense |
Chr. - 12838841 - 12868145 on Build GRCm38 |
View Full Product Details