The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016972.2 | NP_058668.1 | 4 - 5 | 1189 | 106 |
GenBank mRNA | AF171668.1 | - | 4 - 5 | 844 | 106 |
BC059004.1 | - | 4 - 5 | 1189 | 106 | |
Y19022.1 | - | 4 - 5 | 856 | 106 |