Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Slc22a17
- Entrez Gene ID
-
59049
- Gene Name
- solute carrier family 22 (organic cation transporter), member 17
- Gene Aliases
-
1700094C23Rik, 24p3R, AU041908, AW555662, BOIT, Boct, mBOCT
- Chromosome Location
-
Chr.14: 54906727 - 54913139 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_021551.4 |
NP_067526.2 |
2 - 3 |
756 |
63 |
GenBank mRNA |
AK007059.1 |
- |
2 - 3 |
701 |
63 |
View Full Product Details