The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168535.1 | NP_001162007.1 | 5 - 6 | 1124 | 71 |
NM_001168536.1 | NP_001162008.1 | 5 - 6 | 1124 | 71 | |
NM_001168537.1 | NP_001162009.1 | 5 - 6 | 1124 | 71 | |
NM_001168538.1 | NP_001162010.1 | 5 - 6 | 1124 | 71 | |
NM_027986.3 | NP_082262.1 | 5 - 6 | 1124 | 71 | |
GenBank mRNA | AK009280.1 | - | 5 - 6 | 1116 | 71 |
AK032600.1 | - | 5 - 6 | 1103 | 71 | |
AK032813.1 | - | 5 - 6 | 1110 | 71 | |
AK038926.1 | - | 5 - 6 | 1104 | 71 | |
AK039148.1 | - | 5 - 6 | 1112 | 71 | |
BC004588.1 | - | 1 - 2 | 139 | 71 | |
BC006901.1 | - | 5 - 6 | 1086 | 71 |