Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trim13
Entrez Gene ID
66597
Gene Name
tripartite motif-containing 13
Gene Aliases
3110001L12Rik, CAR, LEU5, RNF77, Rfp2
Chromosome Location
Chr.14: 61598226 - 61605946 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164220.1 NP_001157692.1 2 - 3 227 103
NM_023233.3 NP_075722.1 2 - 3 165 103
GenBank mRNA AF220129.1 - 2 - 3 242 103
AK013959.2 - 2 - 3 137 103
AK017850.1 - 1 - 2 122 103
AK162129.1 - 2 - 3 215 103
BC138576.1 - 1 - 2 91 103
BC145915.1 - 1 - 2 91 103
BM933374.1 - 2 - 3 165 103

View Full Product Details