Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Batf3
- Entrez Gene ID
-
381319
- Gene Name
- basic leucine zipper transcription factor, ATF-like 3
- Gene Aliases
-
9130211I03Rik, Snft
- Chromosome Location
-
Chr.1: 191098414 - 191108943 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK171970.1 |
- |
- |
513 |
67 |
View Full Product Details