Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Depdc1a
Entrez Gene ID
76131
Gene Name
DEP domain containing 1a
Gene Aliases
5830484J08Rik, Depdc1
Chromosome Location
Chr.3: 159495426 - 159529955 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172092.1 NP_001165563.1 3 - 4 554 166
NM_001172093.1 NP_001165564.1 3 - 4 554 166
NM_029523.3 NP_083799.2 3 - 4 554 166
GenBank mRNA AK050456.1 - 3 - 4 491 166
AK088719.1 - 3 - 4 530 166
AK146458.1 - 3 - 4 525 166
BC023934.1 - 3 - 4 547 166

View Full Product Details