Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Depdc1a
Entrez Gene ID
76131
Gene Name
DEP domain containing 1a
Gene Aliases
5830484J08Rik, Depdc1
Chromosome Location
Chr.3: 159495426 - 159529955 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172092.1 NP_001165563.1 11 - 12 2367 139
NM_001172093.1 NP_001165564.1 10 - 11 1536 139
GenBank mRNA AK050456.1 - 10 - 11 1473 139
AK088719.1 - 10 - 11 1512 139
AK146458.1 - 11 - 12 2336 139
BC005799.1 - 4 - 5 690 139

View Full Product Details