The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278218.1 | NP_001265147.1 | 1 - 2 | 207 | 59 |
GenBank mRNA | BC043102.1 | - | 1 - 2 | 168 | 59 |
BC059225.1 | - | 1 - 2 | 168 | 59 |