The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029145.3 | NP_083421.2 | 1 - 2 | 146 | 64 |
GenBank mRNA | AY226990.1 | - | 1 - 2 | 146 | 64 |
BC141116.1 | - | 1 - 2 | 139 | 64 |