Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sdccag8
Entrez Gene ID
76816
Gene Name
serologically defined colon cancer antigen 8
Gene Aliases
2700048G21Rik, 5730470G24Rik, Cccap
Chromosome Location
Chr.1: 176814526 - 177020437 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029756.3 NP_084032.1 4 - 5 749 143
GenBank mRNA AK012392.1 - 4 - 5 537 143
AK017691.1 - 4 - 5 532 143
AK051683.1 - 4 - 5 552 143
AK132316.1 - 4 - 5 545 143
AK133622.1 - 4 - 5 493 143
AK136921.1 - 4 - 5 572 143
BC016244.1 - 4 - 5 529 143

View Full Product Details