The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027839.2 | NP_082115.2 | 1 - 2 | 130 | 84 |
GenBank mRNA | AK018613.1 | - | 1 - 2 | 127 | 84 |
AK033581.2 | - | 1 - 2 | 131 | 84 | |
BC049361.1 | - | 1 - 2 | 111 | 84 |