The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027808.1 | NP_082084.1 | 13 - 14 | 3972 | 118 |
GenBank mRNA | AK173191.1 | - | 12 - 13 | 5684 | 118 |