Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Furin
Entrez Gene ID
18550
Gene Name
furin (paired basic amino acid cleaving enzyme)
Gene Aliases
9130404I01Rik, Fur, PACE, Pcsk3, SPC1
Chromosome Location
Chr.7: 80389194 - 80405445 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081454.1 NP_001074923.1 15 - 16 2066 87
NM_011046.2 NP_035176.1 15 - 16 2186 87
GenBank mRNA AK040581.1 - - 3030 87
BC048234.1 - 14 - 15 1893 87
L26489.1 - 15 - 16 2186 87
X54056.1 - 14 - 15 1889 87

View Full Product Details