Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Slc9b1
- Entrez Gene ID
-
74446
- Gene Name
- solute carrier family 9, subfamily B (NHA1, cation proton antiporter 1), member 1
- Gene Aliases
-
1700094G20Rik, 4933424B12Rik, 4933425K02Rik, Nhedc1, mtsNHE
- Chromosome Location
-
Chr.3: 135347994 - 135397827 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK016917.1 |
- |
9 - 10 |
1597 |
87 |
View Full Product Details