Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ska3
Entrez Gene ID
219114
Gene Name
spindle and kinetochore associated complex subunit 3
Gene Aliases
F630043A04Rik
Chromosome Location
Chr.14: 57806560 - 57826190 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198605.3 NP_941007.1 3 - 4 385 94
GenBank mRNA AK089223.1 - 3 - 4 386 94
AK158142.1 - 3 - 4 407 94
BC117500.1 - 3 - 4 377 94
BC117501.1 - 3 - 4 377 94

View Full Product Details