Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Htr2a
Entrez Gene ID
15558
Gene Name
5-hydroxytryptamine (serotonin) receptor 2A
Gene Aliases
E030013E04, Htr-2, Htr2
Chromosome Location
Chr.14: 74640840 - 74706859 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172812.2 NP_766400.1 2 - 3 1705 79
GenBank mRNA AK053140.1 - 2 - 3 1705 79
BC108972.1 - 2 - 3 734 79
BC108973.1 - 2 - 3 734 79
S49542.1 - 2 - 3 612 79

View Full Product Details