Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccnb1ip1
Entrez Gene ID
239083
Gene Name
cyclin B1 interacting protein 1
Gene Aliases
Gm288, Hei10, mei4
Chromosome Location
Chr.14: 50789249 - 50795983 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111119.1 NP_001104589.1 3 - 4 745 87

View Full Product Details