Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Agbl1
Entrez Gene ID
244071
Gene Name
ATP/GTP binding protein-like 1
Gene Aliases
Ccp4, D430020F16, EG244071, Nna1-l1
Chromosome Location
Chr.7: 76229887 - 77124698 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199224.1 NP_001186153.1 8 - 9 900 63
GenBank mRNA AK052433.1 - 2 - 3 386 63
DQ867033.1 - 2 - 3 386 63
FN429927.1 - 8 - 9 900 63

View Full Product Details